missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPR88 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB108281-100
This item is not returnable.
View return policy
Description
GPR88 Monoclonal antibody specifically detects GPR88 in Human samples. It is validated for Immunocytochemistry
Specifications
| GPR88 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 54112 | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 1039818 | |
| Immunocytochemistry 3-25 ug/mL | |
| G protein coupled receptor 88, G protein-coupled receptor 88, G-protein coupled receptor 88, probable G-protein coupled receptor 88, STRG, Striatum-specific G-protein coupled receptor | |
| Human GPR88 synthetic peptide | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction