missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPR56 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB4636R-100UG
This item is not returnable.
View return policy
Description
GPR56 Monoclonal specifically detects GPR56 in Human samples. It is validated for Flow Cytometry.
Specifications
| GPR56 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| BFPP, EGF-TM7-like, G protein-coupled receptor 56,7-transmembrane protein with no EGF-like N-terminal domains-1, Protein TM7XN1, TM7LN4, TM7LN4G-protein coupled receptor 56, TM7XN1, TM7XN1DKFZp781L1398 | |
| Chinese hamster ovary cell line CHO-derived recombinant human GPR56, Met1-Val342, Accession No. Q9Y653 | |
| 100 μg | |
| 9289.0 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 776123 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| Q9Y653 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human GPR56 in direct ELISA. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction