missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPR37L1 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC4449T-100UG
This item is not returnable.
View return policy
Description
GPR37L1 Monoclonal specifically detects GPR37L1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| GPR37L1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| O60883 | |
| GPR37L1 | |
| NS0 mouse myeloma cell line transfected with human GPR37L1 Accession # O60883 | |
| 100 μg | |
| Primary | |
| Detects human GPR37L1. Stains human GPR37L1 transfectants but not irrelevant transfectants. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry | |
| 444532 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| endothelin B receptor-like protein 2, ET(B)R-LP-2, ETBRLP2, ETBR-LP-2, ETBR-LP-2endothelin type b receptor-like protein 2, G protein-coupled receptor 37 like 1, G-protein coupled receptor 37 like 1, G-protein coupled receptor 37-like 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9283 | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction