missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPR26 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB8449V-100UG
This item is not returnable.
View return policy
Description
GPR26 Monoclonal specifically detects GPR26 in Human samples. It is validated for Flow Cytometry.
Specifications
| GPR26 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q8NDV2 | |
| GPR26 | |
| NS0 mouse myeloma cell line transfected with human GPR26 Met1-Glu337 Accession # Q8NDV2 | |
| RUO | |
| 2849 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 891318 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| DKFZp761H2121, FLJ33774, G protein-coupled receptor 26, G-protein coupled receptor 26, MGC138216 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Stains human GPR26 transfectants but not irrelevant transfectants in flow cytometry. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction