missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GPR18 (aa 161-192) Control Fragment Recombinant Protein

Product Code. 30200745
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30200745

Brand: Invitrogen™ RP90882

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. GPR18 is an orphan G protein-coupled receptor found at high levels in the testis, small intestines and cells associated with the immune system including lymphocytes, thymus and spleen. GPR18 has been found to be a receptor for endogenous lipid neurotransmitters, several of which also bind to cannabinoid receptors. Some ligands described to be GPR18 agonists or partial agonists include N-Arachidonoylglycine, Abnormal cannabidiol, O-1602, AM-251, Cannabidiol, delta 9-Tetrahydrocannabinol, N-arachidonoyl ethanolamine and Arachidonylcyclopropylamide. Antagonists include the synthetic cannabidiol related compounds CID-85469571 and O-1918.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14330
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 2841
Name Human GPR18 (aa 161-192) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias EMBL:AAI28783.1}; G protein-coupled receptor 18; GPCRW; Gpr18; gpr18 {ECO:0000312; G-protein coupled receptor 18; NAGly receptor; N-arachidonyl glycine receptor; RP11-461N23.8
Common Name GPR18
Gene Symbol GPR18
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KDPDKDSTPATCLKISDIIYLKAVNVLNLTRL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.