missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GPR150 (aa 358-434) Control Fragment Recombinant Protein

Product Code. 30200044
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30200044

Brand: Invitrogen™ RP89583

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (69%), Rat (69%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-110920 (PA5-110920. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

G protein-coupled receptors (GPCRs), also designated seven transmembrane (7TM) receptors and heptahelical receptors, are a protein family which interact with G proteins (heterotrimeric GTPases) to synthesize intracellular second messengers such as diacylglycerol, cyclic AMP, inositol phosphates, and calcium ions. Their diverse biological functions range from vision and olfaction to neuronal and endocrine signaling and are involved in many pathological conditions. G protein receptor 150 (GPR150), also designated PGR11 and seven transmembrane helix receptor, is a member of the rhodopsin family of GCPRs and is involved in signal transduction. GPR150 is expressed in various human tissues, including normal small intestine, skeletal muscle, kidney and tonsil, as well as cancerous blood, bladder, placenta and parathyroid.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NGU9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 285601
Name Human GPR150 (aa 358-434) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C030001A19Rik; G protein-coupled receptor 150; Gpr150; G-protein coupled receptor PGR11; GPRv40; Pgr11; probable G-protein coupled receptor 150; RGD1564729; seven transmembrane helix receptor
Common Name GPR150
Gene Symbol GPR150
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GDCRLRRQLRKRLGSLCCAPQGGAEDEEGPRGHQALYRQRWPHPHYHHARREPLDEGGLRPPPPRPRPLPCSCESAF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.