missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GPR126 (aa 467-576) Control Fragment Recombinant Protein

Product Code. 30200849
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30200849

Brand: Invitrogen™ RP88750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (55%), Rat (55%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

GPR126 encodes a product that is upregulated in human umbilical vein endothelial cells, encodes a G protein-coupled receptor. Variations in this gene can affect a persoNULLs stature. Multiple transcript variants encoding different proteins have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q86SQ4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 57211
Name Human GPR126 (aa 467-576) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1190004A11Rik; ADGRG6; ADGRG6 C-terminal fragment; ADGRG6 N-terminal fragment; ADGRG6-CTF; ADGRG6-NTF; adhesion G protein-coupled receptor G6; adhesion G-protein coupled receptor G6; AI449247; APG1; AW045736; bge; big ears; developmentally regulated G-protein-coupled receptor; DREG; G protein-coupled receptor 126; Gm222; gpr126; G-protein coupled receptor 126; HBV PreS1-transactivated protein 2; lau; lauscher; LCCS9; probable G-protein coupled receptor 126; PS1TP2; si:ch211-202p1.3; unm st49; unm_st49; vascular inducible G protein-coupled receptor; vascular-inducible G protein-coupled receptor; VIGR
Common Name GPR126
Gene Symbol ADGRG6
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KRSLEDEPRLVLWALLVYNATNNTNLEGKIIQQKLLKNNESLDEGLRLHTVNVRQLGHCLAMEEPKGYYWPSIQPSEYVLPCPDKPGFSASRICFYNATNPLVTYWGPVD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.