missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPR12 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB6738T-100UG
This item is not returnable.
View return policy
Description
GPR12 Monoclonal specifically detects GPR12 in Human samples. It is validated for Flow Cytometry.
Specifications
| GPR12 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. | |
| FLJ18149, FLJ97704, G protein-coupled receptor 12, GPCR12, GPCR21, G-protein coupled receptor 12, MGC138349, MGC138351 | |
| NS0 mouse myeloma cell line transfected with human GPR12 Met1-Val334 Accession # P47775 | |
| 100 μg | |
| 2835 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 584940 | |
| Flow Cytometry 0.25-1μg/106 cells | |
| P47775 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human GPR12 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction