missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPR116 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB112481S-100UG
This item is not returnable.
View return policy
Description
GPR116 Monoclonal antibody specifically detects GPR116 in Human samples. It is validated for Flow Cytometry
Specifications
| GPR116 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| FLJ90640, G protein-coupled receptor 116, Ig-Hepta homolog, KIAA0758DKFZp564O1923, KPG_001, probable G-protein coupled receptor 116 | |
| E. coli -expressed recombinant Human GPR116 extracellular domain., His643-Asn945, Accession # Q8IZF2 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 1055815 | |
| Flow Cytometry | |
| Q8IZF2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 221395 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction