missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPR111 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB64941-100
This item is not returnable.
View return policy
Description
GPR111 Monoclonal specifically detects GPR111 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| GPR111 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| G protein-coupled receptor 111, PGR20 | |
| Chinese Hamster Ovary cell line, CHO-derived human GPR111, Cys19-Lys375, Accession # Q8IZF7 | |
| 100 μg | |
| Primary | |
| Detects human GPR111 in direct ELISAs. Stains human GPR111 HEK293-transfected cells and does not stain irrelevant non-GPR111 transfectants by Flow Cytometry. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 594522 | |
| Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q8IZF7 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 222611 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction