missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPR111 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£143.00 - £388.00
Specifications
| Antigen | GPR111 |
|---|---|
| Clone | 594519 |
| Dilution | Flow Cytometry 0.25 μg/106 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18724397
![]() |
R&D Systems
MAB6494-SP |
25 μg |
£143.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18772707
![]() |
R&D Systems
MAB6494-100 |
100 μg |
£388.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPR111 Monoclonal specifically detects GPR111 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| GPR111 | |
| Flow Cytometry 0.25 μg/106 cells, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| G protein-coupled receptor 111, PGR20 | |
| Chinese Hamster Ovary cell line, CHO-derived recombinant human GPR111, Cys19-Lys375, Accession # Q8IZF7 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human GPR111 in direct ELISAs. |
| 594519 | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Human | |
| Q8IZF7 | |
| 222611 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title