missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GPD2 Recombinant Protein

Product Code. 11528981
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11528981 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11528981 Supplier Invitrogen™ Supplier No. RP75656

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This product contains purified recombinant human mitochondiral Glycerol-3-PO4 Dehydrogenase protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.

Mitochondrial glycerophosphate dehydrogenase (EC 1. 1. 99. 5), or GPD2, is located on the outer surface of the inner mitochondrial membrane and catalyzes the unidirectional conversion of glycerol-3-phosphate (G-3-P) to dihydroxyacetone phosphate (DHAP) with concomitant reduction of the enzyme-bound FAD. Together with a cytosolic NAD-linked GPD (GPD1; MIM 138420), GPD2 forms the glycerol phosphate shuttle, which uses the interconversion of G-3-P and DHAP to transfer reducing equivalents into mitochondria, resulting in the reoxidation of NADH formed during glycolysis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P43304
Concentration 0.095 mg/mL
For Use With (Application) Control, Western Blot
Formulation 20 mM tris HCl with 1 mM EDTA, 50% glycerol and no preservative; pH 8
Gene ID (Entrez) 2820
Name Human GPD2
Quantity 10 μg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias AA408484; AI448216; AU021455; AW494132; FAD-linked glycerol-3-phosphate dehydrogenase; GDH2; Gdm1; glycerol phosphate dehydrogenase 1, mitochondrial; glycerol phosphate dehydrogenase 2, mitochondrial; Glycerol-3-phosophate dehydrogenase 2 (mitochondrial); glycerol-3-phosphate dehydrogenase 2; glycerol-3-phosphate dehydrogenase 2 (mitochondrial); glycerol-3-phosphate dehydrogenase 2, mitochondrial; glycerol-3-phosphate dehydrogenase, mitochondrial; Gpd2; GPDH; GPDH-M; GPDM; GPD-M; mGPDH; mitochondrial glycerol-3-phosphate dehydrogenase; mitochondrial glycerophosphate dehydrogenase; mtGPD; mtGPDH; mtGPDH gene, promoter region and alternative transcripts; Protein TISP38; testicular tissue protein Li 76; TISP38
Common Name GPD2
Gene Symbol GPD2
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Ala43-Tyr600 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.