missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GPD2 (aa 547-664) Control Fragment Recombinant Protein

Product Code. 30209935
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30209935

Brand: Invitrogen™ RP102982

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61022 (PA5-61022. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Mitochondrial glycerophosphate dehydrogenase (EC 1. 1. 99. 5), or GPD2, is located on the outer surface of the inner mitochondrial membrane and catalyzes the unidirectional conversion of glycerol-3-phosphate (G-3-P) to dihydroxyacetone phosphate (DHAP) with concomitant reduction of the enzyme-bound FAD. Together with a cytosolic NAD-linked GPD (GPD1; MIM 138420), GPD2 forms the glycerol phosphate shuttle, which uses the interconversion of G-3-P and DHAP to transfer reducing equivalents into mitochondria, resulting in the reoxidation of NADH formed during glycolysis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P43304
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 2820
Name Human GPD2 (aa 547-664) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA408484; AI448216; AU021455; AW494132; FAD-linked glycerol-3-phosphate dehydrogenase; GDH2; Gdm1; glycerol phosphate dehydrogenase 1, mitochondrial; glycerol phosphate dehydrogenase 2, mitochondrial; Glycerol-3-phosophate dehydrogenase 2 (mitochondrial); glycerol-3-phosphate dehydrogenase 2; glycerol-3-phosphate dehydrogenase 2 (mitochondrial); glycerol-3-phosphate dehydrogenase 2, mitochondrial; glycerol-3-phosphate dehydrogenase, mitochondrial; Gpd2; GPDH; GPDH-M; GPDM; GPD-M; mGPDH; mitochondrial glycerol-3-phosphate dehydrogenase; mitochondrial glycerophosphate dehydrogenase; mtGPD; mtGPDH; mtGPDH gene, promoter region and alternative transcripts; Protein TISP38; testicular tissue protein Li 76; TISP38
Common Name GPD2
Gene Symbol Gpd2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TAVDMISRRTRLAFLNVQAAEEALPRIVELMGRELNWDDYKKQEQLETARKFLYYEMGYKSRSEQLTDRSEISLLPSDIDRYKKRFHKFDADQKGFITIVDVQRVLESINVQMDENTL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.