missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human gp130 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB628-500
This item is not returnable.
View return policy
Description
gp130/CD130 Monoclonal specifically detects gp130/CD130 in Human samples. It is validated for Immunohistochemistry, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| gp130/CD130 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, IL6ST, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3572 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Immunohistochemistry, Neutralization, ELISA | |
| 28105 | |
| Immunohistochemistry 5-25 ug/mL, Neutralization 0.02-0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P40189 | |
| IL6ST | |
| Sf 21-derived recombinant human gp130 extracellular domain Leu24-Glu619 (Glu619-Asp, predicted) Accession # P40189 | |
| 500 μg | |
| Primary | |
| Detects human gp130 in ELISAs. In sandwich immunoassays, no significant cross-reactivity or interference with recombinant human (rh) CNTF, rhIL-6, rhIL-6 R, recombinant mouse (rm) IL-6, rhIL-11, rhLIF, rhLIF R, rmLIF, or rhOSM is observed. This antibody blocks the human gp130-mediated bioactivities induced by IL-6, IL-11, LIF, OSM, and CNTF. | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction