missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB3B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20045
This item is not returnable.
View return policy
Description
RAB3B Polyclonal specifically detects RAB3B in Human, Mouse samples. It is validated for Western Blot.
Specifications
| RAB3B | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| brain antigen RAB3B, RAB3B, member RAS oncogene family, ras-related protein Rab-3B | |
| Rabbit | |
| 25 kDa | |
| 0.1 mL | |
| Signal Transduction | |
| 5865 | |
| Human | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| P20337 | |
| RAB3B | |
| Recombinant protein encompassing a sequence within the center region of human RAB3B. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Mouse (98%), Rat (97%), Bovine (98%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction