missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GOLGA4 (aa 1290-1380) Control Fragment Recombinant Protein

Product Code. 30195350
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195350

Brand: Invitrogen™ RP108831

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (80%), Rat (80%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. This gene encodes one of the golgins, a family of proteins localized to the Golgi. This protein has been postulated to play a role in Rab6-regulated membrane-tethering events in the Golgi apparatus. Alternatively spliced transcript variants encoding different isoforms have been identified in this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13439
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 2803
Name Human GOLGA4 (aa 1290-1380) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 256 kDa golgin; 72.1 protein; AI225887; AU019508; centrosome-related protein F46; CRPF46; EMBL:AAH85124.1, ECO:0000312; GCP2; GOLG; Golga4; golga4 {ECO:0000312; golgi autoantigen, golgin subfamily a, 4; golgin A4; Golgin subfamily A member 4; golgin subfamily A member 4 {ECO:0000250; golgin-240; golgin-245; MU-RMS-40.18; Olp-1; p230; protein 72.1; RGD:1591925}; tGolgin-1; trans-Golgi p230; UniProtKB:Q13439}
Common Name GOLGA4
Gene Symbol GOLGA4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ISFQQATHQLEEKENQIKSMKADIESLVTEKEALQKEGGNQQQAASEKESCITQLKKELSENINAVTLMKEELKEKKVEISSLSKQLTDLN
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.