missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GnRHR (aa 1-38) Control Fragment Recombinant Protein

Product Code. 30204435
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30204435

Brand: Invitrogen™ RP105725

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-110782 (PA5-110782. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes the receptor for type 1 gonadotropin-releasing hormone. This receptor is a member of the seven-transmembrane, G-protein coupled receptor (GPCR) family. It is expressed on the surface of pituitary gonadotrope cells as well as lymphocytes, breast, ovary, and prostate. Following binding of gonadotropin-releasing hormone, the receptor associates with G-proteins that activate a phosphatidylinositol-calcium second messenger system. Activation of the receptor ultimately causes the release of gonadotropic luteinizing hormone (LH) and follicle stimulating hormone (FSH). Defects in this gene are a cause of hypogonadotropic hypogonadism (HH). Alternative splicing results in multiple transcript variants encoding different isoforms. More than 18 transcription initiation sites in the 5 region and multiple polyA signals in the 3 region have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P30968
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 2798
Name Human GnRHR (aa 1-38) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias GH1; GnRH receptor; Gnrhr; gnRH-R; GNRHR1; gonadotropin releasing hormone receptor; gonadotropin releasing hormone receptor type-I; gonadotropin-releasing hormone (type 1) receptor 1; gonadotropin-releasing hormone receptor; GRHR; growth hormone; HH7; HHG; hypergonadotropic hypogonadism; LCGR; leutinizing hormone releasing horomone receptor; leutinizing-releasing hormone receptor; LGR2; LH/CGR; LH/CG-R; LHCGR; LHR; Lh-rh receptor; Lhrhr; LRHR; LSH-R; luliberin receptor; Luteinizing hormone receptor; luteinizing hormone/choriogonadotropin receptor; lutropin/choriogonadotropin receptor; lutropin-choriogonadotropic hormone receptor; type I GnRH receptor; Type I GNRHR1; ULG5
Common Name GnRHR
Gene Symbol Gnrhr
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MANSASPEQNQNHCSAINNSIPLMQGNLPTLTLSGKIR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.