missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Luteinizing Hormone Releasing Hormone Synthetic Peptide, PEP-10933, from Invitrogen.
The Luteinizing Hormone Releasing Hormone (LH-RH) is produced in the hypothalamus and influences the release of Luteinizing Hormone in the pituitary gland. LH-RH is also known as Gonadotropin-releasing hormone (Gn-RH).
Specifications
Specifications
| Gene ID (Entrez) | 2796 |
| Species | Human |
| Purification Method | purified |
| Accession Number | P01148 |
| Common Name | Human GnRH1 |
| Content And Storage | -20°C. Avoid freeze-thaw cycles. |
| Description | Synthetic Peptide |
| Format | Lyophilized |
| Formulation | peptide with no preservative |
| For Use With (Application) | Control |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?