missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GNAL (aa 111-152) Control Fragment Recombinant Protein

Product Code. 30208392
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30208392

Brand: Invitrogen™ RP108391

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84051 (PA5-84051. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Guanine nucleotide-binding proteins (G proteins) are involved as modulators or transducers in various transmembrane signaling systems. G(olf) alpha mediates signal transduction within the olfactory neuroepithelium and the basal ganglia. May be involved in some aspect of visual transduction, and in mediating the effect of one or more hormones/neurotransmitters.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P38405
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 2774
Name Human GNAL (aa 111-152) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2610011C15Rik; 9630020G10Rik; adenylate cyclase-stimulating G alpha protein, olfactory type; DYT25; G alpha 10; G protein subunit alpha L; Galphaolf; Gna10; Gnal; Golf; guanine nucleotide binding protein (G protein), alpha activating activity polypeptide, olfactory type; guanine nucleotide binding protein (G protein), alpha stimulating activity polypeptide, olfactory type; guanine nucleotide binding protein, alpha 10; guanine nucleotide binding protein, alpha stimulating, olfactory type; guanine nucleotide-binding protein G(olf) subunit alpha; guanine nucleotide-binding protein G(olf), alpha subunit; Olf; olfa; Olf-alpha protein (olfactory neuron-specific G protein); RGD1305940
Common Name GNAL
Gene Symbol Gnal
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NQFRSDYIKSIAPITDFEYSQEFFDHVKKLWDDEGVKACFER
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.