missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human GNAI1/GNAI2 (aa 346-355) Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Human G-protein alpha Inhibitor (residues 346-355) Synthetic Peptide, PEP-129, from Invitrogen.
The G protein family of signal transducers includes the alpha, beta, gamma subunits and the effector which are most clearly distinguished by their different alpha chains. The family also includes Gs and Gi, the stimulatory and inhibitory GTP-binding regulators of adenylate cyclase; Go, a protein of unknown function that is abundantly expressed in brain; and transducin 1 and 2, proteins involved in retinal phototransduction. The G proteins possess GTPase activity and are components of a complex membrane signaling system that includes membrane-bound receptors. The system exists to modulate extracellular signals as they are transmitted into the cell.
Specifications
Specifications
| Gene ID (Entrez) | 2770,2771 |
| Species | Human |
| Purification Method | purified |
| Accession Number | P04899,P63096 |
| Common Name | Human GNAI1/GNAI2 (aa 346-355) |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C. Avoid freeze-thaw cycles. |
| Description | Synthetic Peptide |
| Format | Lyophilized |
| Formulation | peptide with no preservative |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction