missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GMF-beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1276-SP
This item is not returnable.
View return policy
Description
GMF-beta Monoclonal specifically detects GMF-beta in Human samples. It is validated for Western Blot.
Specifications
| GMF-beta | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P60983 | |
| GMFB | |
| E. coli-derived recombinant human GMF-beta Met1-His142 Accession # P60983 | |
| 25 μg | |
| Primary | |
| Detects human GMF-beta in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 185003 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| glia maturation factor beta, glia maturation factor, beta, GMF, GMFB, GMFbeta, GMF-beta | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2764 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction