missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GM2/GD2 Synthase/ B4GALNT1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £417.00
Specifications
| Antigen | B4GALNT1 |
|---|---|
| Clone | 1024912 |
| Dilution | Immunohistochemistry 5-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18751487
![]() |
R&D Systems
MAB10503-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18752917
![]() |
R&D Systems
MAB10503-100 |
100 μg |
£417.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
B4GALNT1 Monoclonal specifically detects B4GALNT1 in Human samples. It is validated for Immunohistochemistry.Specifications
| B4GALNT1 | |
| Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| (N-acetylneuraminyl)-galactosylglucosylceramide, beta-1,4 N-acetylgalactosaminyltransferase 1, beta1,4GalNAc-T, beta-1,4-N-acetyl-galactosaminyl transferase 1, beta1-4GalNAc-T, GALGT, GALGTEC 2.4.1.92, GalNAc-T, GALNACT, GD2 Synthase, GD2 synthase, GM2 synthase, GM2 Synthase, GM2/GD2 synthase, SIAT2, SPG26, UDP-Gal:betaGlcNAc beta-1,4-N-acetylgalactosaminyltransferase transferase 1, UDP-N-acetyl-alpha-D-galactosamine:(N-acetylneuraminyl)-galactosylglucosylceramideN-acetylgalactosaminyltransferase (GalNAc-T) | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human GM2/GD2 Synthase/ B4GALNT1, Ser26-Gln533, Accession # Q00973 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1024912 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q00973 | |
| 2583 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title