missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GM-CSF R alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
£177.00 - £460.00
Specifications
| Antigen | GM-CSFR alpha |
|---|---|
| Clone | 31909 |
| Dilution | Immunohistochemistry 5-15 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18122501
|
R&D Systems
MAB7064-100 |
100 μg |
£460.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18120501
![]() |
R&D Systems
MAB7064-SP |
25 μg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GM-CSFR alpha Monoclonal antibody specifically detects GM-CSFR alpha in Human samples. It is validated for ImmunohistochemistrySpecifications
| GM-CSFR alpha | |
| Immunohistochemistry 5-15 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 1438 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
| 31909 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD116, CD116 antigen, CDw116, colony stimulating factor 2 receptor alpha subunit, colony stimulating factor 2 receptor, alpha, low-affinity(granulocyte-macrophage), CSF2R, CSF2RA, CSF2RAX, CSF2RAY, CSF2RX, CSF2RY, GM-CSF receptor alpha subunit, GMCSFR, GMCSFR alpha, GM-CSFRa, GM-CSF-R-alpha, GMCSFR-alpha, GMR, GMR-alpha, granulocyte-macrophage colony-stimulating factor receptor alpha chain, granulocyte-macrophage colony-stimulating factor receptor subunit alpha, MGC3848, MGC4838 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human GM-CSF R alpha , Extracellular domain | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title