missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GM-CSF Human Uncoated ELISA Kit with Plates
Specifically engineered for accurate and precise measurement protein levels from samples including serum, plasma, and cell culture supernatants. This second generation kit has increased sensitivity, with a standard curve range from 6-750pg/mL.
£174.00 - £524.00
Specifications
Target Species | Human |
---|---|
For Use With (Application) | ELISA |
Synonym | Granulocyte Macrophage Colony-Stimulating Factor, CSF2 |
Alert:
Contains Sodium azide
Product Code | Brand | Quantity | Gene Symbol | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Gene Symbol | Price | Quantity & Availability | ||||
15581217
|
Invitrogen™
88-8337-22 |
2 x 96 Tests | CLMF,CLMF2,IL-12B,IMD28,IMD29,NKSF,NKSF2,CLMF,IL-12A,NFSK,NKSF1,P35 |
£174.00
Pack of 2 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
15511227
|
Invitrogen™
88-8337-88 |
10 x 96 Tests | IL-1,IL1-BETA,IL1F2 |
£367.00
Pack of 10 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
15501227
|
Invitrogen™
88-8337-86 |
10 x 96 Tests | CLMF,CLMF2,IL-12B,IMD28,IMD29,NKSF,NKSF2,CLMF,IL-12A,NFSK,NKSF1,P35 |
£392.00
Pack of 10 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
15591217
|
Invitrogen™
88-8337-77 |
20 x 96 Tests | CSF, GM-CSF |
£524.00
Pack of 20 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
Description
Human GM-CSF Ready-SET-Go! (2nd Generation) ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)Specifications
Human | |
ELISA | |
Granulocyte Macrophage Colony-Stimulating Factor, CSF2 |
For Research Use Only