missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GM-CSF Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 19 publications
Brand: R&D Systems MAB215-100
This item is not returnable.
View return policy
Description
GM-CSF Monoclonal specifically detects GM-CSF in Human samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.
Specifications
| GM-CSF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1437 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Neutralization, Immunocytochemistry | |
| 3209 | |
| Western Blot 1 ug/mL, Neutralization 0.075-0.45 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P04141 | |
| CSF2 | |
| E. coli-derived recombinant human GM-CSF Ala18-Glu144 Accession # P04141 | |
| 100 μg | |
| Primary | |
| Detects human GM-CSF in Western blots. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction