missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Glypican 2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£171.00 - £377.00
Specifications
| Antigen | Glypican 2 |
|---|---|
| Clone | 298402 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18726038
![]() |
R&D Systems
MAB2304-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18119250
|
R&D Systems
MAB2304 |
100 μg |
£377.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glypican 2 Monoclonal specifically detects Glypican 2 in Human samples. It is validated for Western Blot.Specifications
| Glypican 2 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q8N158 | |
| 221914 | |
| Mouse myeloma cell line NS0-derived recombinant human Glypican 2 Pro18-Ser553 Accession # Q8N158 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Glypican 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Glypican 3, 5, 6, or recombinant mouse Glypican 2 is observed. |
| 298402 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 2410016G05Rik, Cerebroglycan, GPC2 | |
| GPC2 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto