missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Glycoprotein V/CD42d Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB42491
This item is not returnable.
View return policy
Description
Glycoprotein V/CD42d Monoclonal specifically detects Glycoprotein V/CD42d in Human samples. It is validated for Western Blot.
Specifications
| Glycoprotein V/CD42d | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| P40197 | |
| GP5 | |
| Mouse myeloma cell line NS0-derived recombinant human Glycoprotein V/CD42d Gln17-Gly523 (predicted) Accession # P40197 | |
| 100 μg | |
| Primary | |
| Detects human Glycoprotein V/CD42d in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) Glycoprotein VI, rhTLR1, or rhTLR2 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 508219 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| platelet glycoprotein V, CD42d, glycoprotein V (platelet), GP5, GPV | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2814 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction