missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Glycophorin A Recombinant Protein

Product Code. 18273265 Shop All Bio Techne Products
Click to view available options
Quantity:
250 μg
50 μg
Unit Size:
250µg
50µg
This item is not returnable. View return policy

Product Code. 18273265

Brand: Novus Biologicals™ NBP259660250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 20-91 of Human Glycophorin A The Recombinant Human Glycophorin A Protein is derived from E. coli. The Recombinant Human Glycophorin A Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) PAGE
Formulation In PBS (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 2993
Molecular Weight (g/mol) 9.1 kDa
Name GYPA Recombinant Protein
Purification Method >85% by SDS-PAGE
Quantity 250 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1μg of protein (determined by LAL method)
Gene Alias CD235a antigen, CD235a, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), Glycophorin A, glycophorin Erik
Gene Symbol GYPA
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.