missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Glycophorin A Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
£167.00 - £1755.00
Specifications
| Antigen | Glycophorin A |
|---|---|
| Clone | R10 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 1.7-25 ug/mL, Immunoprecipitation, CyTOF-ready |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18792749
![]() |
R&D Systems
MAB1228-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18740757
|
R&D Systems
MAB1228-100 |
100 μg |
£437.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18744567
![]() |
R&D Systems
MAB1228-500 |
500 μg |
£1755.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glycophorin A Monoclonal specifically detects Glycophorin A in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF-ready.Specifications
| Glycophorin A | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 1.7-25 ug/mL, Immunoprecipitation, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| CD235a, CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, GYPA, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR | |
| GYPA | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| R10 | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 2993 | |
| Abelson mouse leukemia virus-induced human pre-B tumor cells | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Glycophorin A, the major sialoglycoprotein expressed on red blood cells and erythroid precursor cells [Greaves, M.F. et al. (1983) Blood 61(4):645]. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title