missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Glut3 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB1415S-100UG
This item is not returnable.
View return policy
Description
Glut3 Monoclonal specifically detects Glut3 in Human samples. It is validated for Flow Cytometry.
Specifications
| Glut3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P11169 | |
| SLC2A3 | |
| NS0 mouse myeloma cell line transfected with human Glut3 Accession # P11169 | |
| RUO | |
| 6515 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 202017 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| FLJ90380, GLUT-3, GLUT3Glucose transporter type 3, brain, SLC2A3, solute carrier family 2 (facilitated glucose transporter), member 3, solute carrier family 2, facilitated glucose transporter member 3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Glut3. Recognizes human Glut3 expression on human Glut3-transfected NS0 cells, but not the NS0 control transfectants. No cross-reactivity was observed with transfectants expressing human Glut1 or human Glut2. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction