missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Glut1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB11601-100
This item is not returnable.
View return policy
Description
Glut1 Monoclonal antibody specifically detects Glut1 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunocytochemistry
Specifications
| Glut1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| Choreoathetosis/Spasticity, Episodic (Paroxysmal Choreoathetosis), CSE, DYT17, DYT18, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, GLUT-1, GLUT1DS, HepG2 glucose transporter, HTLVR, Human T-Cell Leukemia Virus (I and II) Receptor, MGC141895, MGC141896, PED, SLC2A1, solute carrier family 2 (facilitated glucose transporter), member 1, Solute Carrier Family 2 Member 1, solute carrier family 2, facilitated glucose transporter member 1 | |
| Synthetic Peptide, Accession # P11166 | |
| 100 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 1084409 | |
| Western Blot 2 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL | |
| P11166 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6513 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction