missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GluR7/GRIK3 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
£206.00 - £449.00
Specifications
| Antigen | GluR7/GRIK3 |
|---|---|
| Clone | 2914A |
| Dilution | Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18192071
|
R&D Systems
MAB113281-SP |
25 μg |
£206.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18181641
|
R&D Systems
MAB113281-100 |
100 μg |
£449.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GluR7/GRIK3 Monoclonal antibody specifically detects GluR7/GRIK3 in Human samples. It is validated for Flow CytometrySpecifications
| GluR7/GRIK3 | |
| Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| EAA5, Excitatory amino acid receptor 5, GLR7, GluK3, gluR-7, GluR7, GluR7a, GLUR7dJ1090M5.1 (glutamate receptor, ionotropic, kainate 3 (GLUR7)), Glutamate receptor 7, glutamate receptor, ionotropic kainate 3, glutamate receptor, ionotropic, kainate 3, GRIK3 | |
| Human embryonic kidney cell, HEK293-derived human GluR7/GRIK3 , Gly249-Met1, Accession # Q13003 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2914A | |
| Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q13003 | |
| 2899 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title