missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GluR6/GRIK2 Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB11512-SP
This item is not returnable.
View return policy
Description
Glutamate Receptor 6 Monoclonal antibody specifically detects Glutamate Receptor 6 in Human samples. It is validated for Western Blot
Specifications
| Glutamate Receptor 6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| EAA4, GLR6, GluK2, GLUK6, glutamate receptor 6, glutamate receptor, ionotropic, kainate 2, ionotropic kainate 2, MGC74427, MRT6 | |
| Chinese Hamster Ovary cell line CHO-derived human GluR6/GRIK2 protein, Thr32-Asn549, Accession # Q13002 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 1060307 | |
| Western Blot 0.5 μg/mL | |
| Q13002 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2898 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction