missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GluR3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB11329-SP
This item is not returnable.
View return policy
Description
Glutamate Receptor 3 Monoclonal antibody specifically detects Glutamate Receptor 3 in Human samples. It is validated for Immunocytochemistry
Specifications
| Glutamate Receptor 3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| AMPA 3, AMPA3, AMPA-selective glutamate receptor 3, GluA3, GluR-3, GLURC, GLUR-C, GLUR-K3, glutamate receptor C, glutamate receptor subunit 3, glutamate receptor, ionotrophic, AMPA 3, GRIA3 | |
| Synthetic Peptide, Accession # P42263 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2b |
| Immunocytochemistry | |
| 1059821 | |
| Immunocytochemistry 5-25 ug/mL | |
| P42263 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2892 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction