missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GluR2 Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
£181.00 - £541.00
Specifications
| Antigen | GluR2 |
|---|---|
| Clone | 1059613 |
| Dilution | Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18786305
![]() |
R&D Systems
MAB11333-100 |
100 μg |
£541.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18726155
![]() |
R&D Systems
MAB11333-SP |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GluR2 Monoclonal antibody specifically detects GluR2 in Human samples. It is validated for Immunohistochemistry, ImmunocytochemistrySpecifications
| GluR2 | |
| Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| AMPA 2, AMPA2, GluA2, GLURB, GluR-K2, glutamate receptor, ionotropic, AMPA 2, GRIA2, HBGR2 | |
| Human GluR2/GRIA2 containing peptide, Accession # P42262 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1059613 | |
| Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P42262 | |
| 2891 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title