missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Glucuronosyltransferase 1A1/UGT1A1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB6490
This item is not returnable.
View return policy
Description
Glucuronosyltransferase 1A1/UGT1A1 Monoclonal specifically detects Glucuronosyltransferase 1A1/UGT1A1 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
Specifications
| Glucuronosyltransferase 1A1/UGT1A1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Simple Western 20 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P22309 | |
| UGT1A1 | |
| E. coli-derived recombinant human Glucuronosyltransferase 1A1/UGT1A1 Leu60-Thr168 Accession # P22309 | |
| 100 μg | |
| Primary | |
| Detects human Glucuronosyltransferase 1A1/UGT1A1 in ELISAs. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 856754 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| bilirubin UDP-glucuronosyltransferase 1-1, bilirubin UDP-glucuronosyltransferase isozyme 1, Bilirubin-specific UDPGT isozyme 1, GNT1, GNT1EC 2.4.1.17, HUG-BR1, UDP glucuronosyltransferase 1 family, polypeptide A1, UDP glycosyltransferase 1 family, polypeptide A1, UDP-glucuronosyltransferase 1-1, UDP-glucuronosyltransferase 1-A, UDP-glucuronosyltransferase 1A1, UDPGT, UDPGT 1-1, UG-BR1, UGT1, UGT1*1, UGT1.1, UGT1-01, UGT1A, UGT-1A, UGT1AUDPGT 1-1, UGT1BILIQTL1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 54658 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction