missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GITR Ligand/TNFSF18 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB6942-SP
This item is not returnable.
View return policy
Description
GITR Ligand/TNFSF18 Monoclonal specifically detects GITR Ligand/TNFSF18 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
Specifications
| GITR Ligand/TNFSF18 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Activation-inducible TNF-related ligand, AITRL, AITRLMGC138237, GITRL, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, TNFSF18, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8995 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF, ELISA | |
| 109114 | |
| Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q9UNG2 | |
| TNFSF18 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human GITR Ligand/TNFSF18 Glu52-Ser177 Accession # Q9UNG2 | |
| 25 μg | |
| Primary | |
| Detects human GITR Ligand/TNFSF18 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) APRIL, rhLIGHT, rhTNF-alpha , or rhVEGI is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction