missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GILT/IFI30 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7715
This item is not returnable.
View return policy
Description
GILT/IFI30 Polyclonal specifically detects GILT/IFI30 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| GILT/IFI30 | |
| Polyclonal | |
| Unconjugated | |
| P13284 | |
| IFI30 | |
| Human embryonic kidney cell line HEK293-derived recombinant human GILT/IFI30 Ser27-Lys250 Accession # P13284 | |
| 100 μg | |
| Primary | |
| Detects human GILT/IFI30 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 1-15 ug/mL | |
| EC 1.8, Gamma-interferon-inducible protein IP-30, GILTgamma-interferon-inducible lysosomal thiol reductase, IFI30, IFI-30, interferon, gamma-inducible protein 30, IP30, IP30interferon gamma-inducible protein 30 preproprotein, Legumaturain, MGC32056 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 10437 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction