missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GFR alpha-3 ELISA Kit

Product Code. 17164503
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
This item is not returnable. View return policy

Product Code. 17164503

Brand: Invitrogen™ EH218RB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human GFR alpha-3 quantitates human GFR alpha-3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human GFR alpha-3.

GFRa-3 binds persephin, thus, persephin, GFRa-3, and Ret PTK form a complex to transduce persephin signal and to mediate persephin function.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number GDNF family receptor alpha 3; GDNF family receptor alpha-3; GDNF receptor alpha-3; GDNF-family receptor alpha 3; GDNFR3; GDNFR-alpha-3; GFR alpha-3; GFRA3; GFRalpha3; GFR-alpha-3; glial cell line derived neurotrophic factor family receptor alpha 3; glial cell line-derived neurotrophic factor receptor alpha-3; GPI-linked receptor; UNQ339/PRO538/PRO3664; Y15110
Assay Range 16.38 to 4,000 pg/mL
Assay Sensitivity 16 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 2676
Gene Symbol Gfra3
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target GFR alpha-3
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.