missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human GFR alpha-2 Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
GFR alpha 2 Synthetic Peptide, PEP-0254, from Invitrogen.
GFRa-2 binds NTN and mediates activation of RET receptor tyrosine kinase by both NTN and GDNF. Thus, NTN, GFRa-2, and the Ret PTK form a complex to transduce NTN signal and to mediate NTN function.
Specifications
Specifications
| Gene ID (Entrez) | 2675 |
| Species | Human |
| Purification Method | purified |
| Accession Number | O00451 |
| Common Name | Human GFR alpha-2 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction