missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GFM2 (aa 661-741) Control Fragment Recombinant Protein

Product Code. 30193577
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30193577

Brand: Invitrogen™ RP96919

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57851 (PA5-57851. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

GFM2 is involved in mitochondrial translation, which is crucial for maintaining mitochondrial function and mutations in this system lead to a breakdown in the respiratory chain-oxidative phosphorylation system and to impaired maintenance of mitochondrial DNA. In Eukaryotes contain two protein translational systems, one in the cytoplasm and one in the mitochondria. This gene encodes one of mitochondrial translation elongation factors. Its role in the regulation of normal mitochondrial function and in different disease states attributed to mitochondrial dysfunction is not known. Alternative splicing results in at least three transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q969S9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 84340
Name Human GFM2 (aa 661-741) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 6530419G12Rik; A930009M04Rik; EFG2; EF-G2mt; EF-G2mt {ECO:0000255; Elongation factor G 2, mitochondrial; elongation factor G 2, mitochondrial {ECO:0000255; Elongation factor G2; G elongation factor; G elongation factor mitochondrial 2; G elongation factor, mitochondrial 2; GFM2; HAMAP-Rule:MF_03059}; hEFG2; mEF-G 2; mEF-G 2 {ECO:0000255; mitochondrial elongation factor G2; mitochondrial ribosome recycling factor 2; MRRF2; MST027; MSTP027; RGD1309854; ribosome-releasing factor 2, mitochondrial; ribosome-releasing factor 2, mitochondrial {ECO:0000255; RRF2; RRF2mt; RRF2mt {ECO:0000255
Common Name GFM2
Gene Symbol Gfm2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TSTTMISACVSRCVQKALKKADKQVLEPLMNLEVTVARDYLSPVLADLAQRRGNIQEIQTRQDNKVVIGFVPLAEIMGYST
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.