missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GFER/ALR Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7940-SP
This item is not returnable.
View return policy
Description
GFER/ALR Monoclonal specifically detects GFER/ALR in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| GFER/ALR | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P55789 | |
| GFER | |
| E. coli-derived recombinant human GFER/ALR Arg82-Asp205 Accession # P55789 | |
| 25 μg | |
| Primary | |
| Detects human GFER/ALR in ELISAs. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 829812 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ALR, ALRFAD-linked sulfhydryl oxidase ALR, Augmenter of liver regeneration, EC 1.8.3.2, ERV1, ERV1 homolog, ERV1hepatopoietin protein, erv1-like growth factor, growth factor, augmenter of liver regeneration, growth factor, erv1 (S. cerevisiae)-like (augmenter of liver regeneration), Hepatopoietin, hERV1, HERV1hepatic regenerative stimulation substance, HPO, HPO1, HPO2, HSS, truncated augmenter of liver regeneration | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2671 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction