missing translation for 'onlineSavingsMsg'
Learn More

Human GDF-3 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18731089
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18731089 100 μg 100µg
18717139 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18731089 Supplier R&D Systems Supplier No. MAB57541

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GDF-3 Monoclonal specifically detects GDF-3 in Human samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDF-3
Applications Neutralization
Classification Monoclonal
Clone 878751
Conjugate Unconjugated
Dilution Neutralization 7.5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9NR23
Gene Alias GDF3, GDF-3, growth differentiation factor 3, growth/differentiation factor 3, KFS3, MCOP7, MCOPCB6, Vgr-2
Gene Symbols GDF3
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived human recombinant GDF-3/BMP-2 heterodimer Ala251-Gly364 Accession # Q9NR23
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9573
Test Specificity Detects human GDF-3 in direct ELISAs. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) BMP-2 and rhGDF-1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.