missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GDF-11/GDF-8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB19582-SP
This item is not returnable.
View return policy
Description
GDF-11/GDF-8 Monoclonal specifically detects GDF-11/GDF-8 in Human samples. It is validated for ELISA, Neutralization.
Specifications
| GDF-11/GDF-8 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GDF11 | |
| E. coli-derived recombinant human GDF-11/BMP-11 Asn299-Ser407 Accession # O95390 | |
| 25 μg | |
| Primary | |
| Detects human GDF-11/BMP-11 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with mouse GDF-8 is observed. | |
| Human | |
| Purified |
| Neutralization | |
| 743835 | |
| Neutralization 0.04-0.2 ug/mL | |
| O95390 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10220 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction