missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human GCP-2 (CXCL6) Recombinant Protein, PeproTech®

Product Code. 17822543
Click to view available options
Quantity:
100 μg
2 x 500 μg
20 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 17822543

Brand: Gibco™ 30041250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

300-41-1MG will be provided as 2 x 500 μg (300-41-500UG). Recombinant Human GCP-2 is a 7.9 kDa protein containing 72 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CXCL6 is a chemotactic for neutrophil granulocytes. It signals through binding and activation of its receptors (CXCR1 and CXCR2). In addition to its chemotactic and angiogenic properties, it has strong antibacterial activity against Gram-positive and Gram-negative bacteria (90-fold-higher when compared to CXCL5 and CXCL7).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P80162
For Use With (Application) Control, Western Blot Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 6372
Molecular Weight (g/mol) 7.9 kDa
Name Human GCP-2 (CXCL6)
Purification Method Purified
Quantity 250 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 5; chemokine (C-X-C motif) ligand 6; chemokine (C-X-C motif) ligand 6 (granulocyte chemotactic protein 2); chemokine alpha 3; CKA3; CKA-3; CXC; CXC chemokine LIX; CXC motif chemokine; C-X-C motif chemokine 5; C-X-C motif chemokine 6; C-X-C motif chemokine ligand 6; CXCL; Cxcl5; CXCL6; Cytokine LIX; GCP2; GCP-2; granulocyte chemotactic protein 2; Scyb5; SCYB6; small inducible cytokine subfamily B (Cys-X-Cys), member 6 (granulocyte chemotactic protein 2); Small inducible cytokine subfamily B (Cys-X-Cys), member b; small-inducible cytokine B5; Small-inducible cytokine B6; Small-inducible cytokine B6, N-processed variant 1; Small-inducible cytokine B6, N-processed variant 2; Small-inducible cytokine B6, N-processed variant 3
Common Name CXCL6
Gene Symbol CXCL6
Biological Activity Determined by its ability to chemoattract human neutrophils using a concentration range of 10.0-50.0 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence VLTELRCTCL RVTLRVNPKT IGKLQVFPAG PQCSKVEVVA SLKNGKQVCL DPEAPFLKKV IQKILDSGNK KN
Content And Storage -20°C
Activity Determined by its ability to chemoattract human neutrophils using a concentration range of 10.0-50.0 ng/ml.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.