missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GCNF/NR6A1 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF3180
This item is not returnable.
View return policy
Description
GCNF/NR6A1 Polyclonal specifically detects GCNF/NR6A1 in Human samples. It is validated for Western Blot.
Specifications
| GCNF/NR6A1 | |
| Polyclonal | |
| Biotin | |
| Q15406 | |
| NR6A1 | |
| E. coli-derived recombinant human GCNF/NR6A1 Met215-Glu480 Accession # Q15406 | |
| 50 μg | |
| Primary | |
| Detects human GCNF/NR6A1 in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| GCNF1, GCNFNR61, Germ cell nuclear factor, hGCNF, NR6A1, nuclear receptor subfamily 6 group A member 1, nuclear receptor subfamily 6, group A, member 1, retinoic acid receptor-related testis-associated receptor, Retinoid receptor-related testis-specific receptor, RTR, RTRhRTR | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2649 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction