missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GCN1L1 (aa 621-748) Control Fragment Recombinant Protein

Product Code. 30198710
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30198710

Brand: Invitrogen™ RP92376

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53848 (PA5-53848. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Acts as a positive activator of the EIF2AK4/GCN2 protein kinase activity in response to amino acid starvation. Forms a complex with EIF2AK4/GCN2 on translating ribosomes; during this process, GCN1 seems to act as a chaperone to facilitate delivery of uncharged tRNAs that enter the A site of ribosomes to the tRNA-binding domain of EIF2AK4/GCN2, and hence stimulating EIF2AK4/GCN2 kinase activity. Participates in the repression of global protein synthesis and in gene-specific mRNA translation activation, such as the transcriptional activator ATF4, by promoting the EIF2AK4/GCN2-mediated phosphorylation of eukaryotic translation initiation factor 2 (eIF-2-alpha/EIF2S1) on 'Ser-52', and hence allowing ATF4-mediated reprogramming of amino acid biosynthetic gene expression to alleviate nutrient depletion. [UniProt]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q92616
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10985
Name Human GCN1L1 (aa 621-748) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4932409G22; AL022764; eIF-2-alpha kinase activator GCN1; G431004K08Rik; Gcn1; GCN1 (general control of amino-acid synthesis 1, yeast)-like 1; GCN1 eIF2 alpha kinase activator homolog; GCN1 eIF-2-alpha kinase activator homolog; GCN1 general control of amino-acid synthesis 1-like 1; GCN1 general control of amino-acid synthesis 1-like 1 (yeast); GCN1, eIF2 alpha kinase activator homolog; GCN1L; GCN1L1; GCN1-like protein 1; General control of amino-acid synthesis 1-like protein 1; hsGCN1; KIAA0219; mKIAA0219; peroxisome proliferator activated receptor interacting complex protein; PRIC295; translational activator GCN1
Common Name GCN1L1
Gene Symbol GCN1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EALVTDAGEVTEAGKAYVPPRVLQEALCVISGVPGLKGDVTDTEQLAQEMLIISHHPSLVAVQSGLWPALLARMKIDPEAFITRHLDQIIPRMTTQSPLNQSSMNAMGSLSVLSPDRVLPQLISTITA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.