missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human GCKR (aa 220-324) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66679 (PA5-66679. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes a protein belonging to the GCKR subfamily of the SIS (Sugar ISomerase) family of proteins. The gene product is a regulatory protein that inhibits glucokinase in liver and pancreatic islet cells by binding non-covalently to form an inactive complex with the enzyme. This gene is considered a susceptibility gene candidate for a form of maturity-onset diabetes of the young (MODY).
Specifications
Specifications
| Accession Number | Q14397 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 2646 |
| Name | Human GCKR (aa 220-324) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | FGQTL5; Gckr; GKRP; GLRE; glucokinase (hexokinase 4) regulator; glucokinase regulator; Glucokinase regulatory protein; hexokinase 4 |
| Common Name | GCKR |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction