missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GATA2 (aa 79-205) Control Fragment Recombinant Protein

Product Code. 30181314
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30181314

Brand: Invitrogen™ RP99376

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

GATA2 (GATA binding protein 2) belongs to the GATA family of transcription factors. GATA proteins play a very important role in signaling pathways controlling proliferation and differentiation of erythroid progenitor cells. There are 6 transcription factors in the GATA family: GATA1 to GATA6. GATA2 is a transcriptional activator regulating endothelin-1 gene expression in endothelial cells. It also appears to be involved in differentiation of other tissue types (uro-genital and nervous) during embryogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P23769
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 2624
Name Human GATA2 (aa 79-205) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias DCML; EMBL:AAK51128.1}; Endothelial transcription factor GATA-2; FLJ45948; GATA binding protein 2; GATA binding protein 3; GATA2; Gata-2; gata2 {ECO:0000312; GATA3; GATA-binding factor 3; GATA-binding protein 2; GATA-bindning protein 2; HDR; HDRS; IMD21; MGC2306; MONOMAC; NFE1B; trans-acting T-cell-specific transcription factor GATA-3; uterine GATA-4 transcription factor
Common Name GATA2
Gene Symbol GATA2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LTGGQMCRPHLLHSPGLPWLDGGKAALSAAAAHHHNPWTVSPFSKTPLHPSAAGGPGGPLSVYPGAGGGSGGGSGSSVASLTPTAAHSGSHLFGFPPTPPKEVSPDPSTTGAASPASSSAGGSAARG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.