missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GATA-6 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF1700
This item is not returnable.
View return policy
Description
GATA-6 Polyclonal specifically detects GATA-6 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| GATA-6 | |
| Polyclonal | |
| Biotin | |
| Q92908-2 | |
| GATA6 | |
| E. coli-derived recombinant human GATA-6 Met1-Ala449 Accession # Q92908-2 | |
| 50 μg | |
| Primary | |
| Detects human GATA-6. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| GATA binding protein 6, GATA6, GATA-binding factor 6, GATA-binding protein 6, transcription factor GATA-6 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2627 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction